missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acetamiprid
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100.0°C to 102.0°C |
| Solubility | Soluble in methanol, chloroform and dimethyl sulfoxide |
| Quantity | 1 g |
| Formula Weight | 222.67 |
| Chemical Name or Material | Acetamiprid |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Fatal if inhaled. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. Store in a well-ventilated place. Keep container tightly closed. |
RUO – Research Use Only