missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ACES, 99%

Catalog No. AAA1155314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1155314 25 g
AAA1155322 100 g
AAA1155336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1155314 Supplier Thermo Scientific Chemicals Supplier No. A1155314
Only null left

CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol

ACES is used as a Good's buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

Solubility
Soluble in water(18g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym aces, n-carbamoylmethyl taurine, n-2-acetamido-2-aminoethanesulfonic acid, 2-2-amino-2-oxoethyl amino ethanesulfonic acid, ethanesulfonic acid, 2-2-amino-2-oxoethyl amino, unii-i5spl5yvby, i5spl5yvby, 2-carbamoylmethylamino ethanesulfonic acid
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Specifications

Melting Point ∼275°C (decomposition)
Quantity 25 g
Beilstein 2253770
Merck Index 14,36
Solubility Information Soluble in water(18g/L).
Formula Weight 182.2
Percent Purity 99%
Chemical Name or Material ACES
EINECSNumber 230-908-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only