missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Ace Chromatography Column with Fritted Disc

Catalog No. 111009321
Change view
Click to view available options
Length (Metric):
46 cm
51 cm
61 cm
Diameter (Metric):
10 mm
19 mm
25 mm
50 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111009321 46 cm 10 mm
111009292 61 cm 19 mm
111009263 51 cm 25 mm
111008984 61 cm 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111009321 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1371621EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No. 15 Ace-Thred at top and 2 mm bore, solid-PTFE stopcock-plug (tapered 1:5) at bottom. Supplied with nylon and FETFE™ O-ring for connecting 12.5 to 14 mm O.D. tube to column, and with tubing connector.

Note: When using tubing connector, O-ring supplied with bushing is NOT necessary. Fritted disc has 70 to 100 μm porosity, sealed into bottom of column.

Specifications

Diameter (Metric) 10 mm
Length (Metric) 46 cm
Product Line Ace
Column Type Chromatography
Material Borosilicate Glass
Technique LPLC