missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Ace Chromatography Column
Description
Note: When using tubing connector, O-ring supplied with bushing is NOT necessary. Fritted disc has 70 to 100 μm porosity, sealed into bottom of column.
Specifications
Specifications
| Diameter (Metric) | 10 mm |
| Length (Metric) | 46 cm |
| Product Line | Ace |
| Column Type | Chromatography |
| Material | Borosilicate Glass |