Learn More
Promega AccuMAP Low pH Protein Digestion Mini Kit, 1 each
Catalog No. PRVA1040
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRVA1040 1 Each
1 options
Catalog No. PRVA1040 Supplier Promega Supplier No. VA1040
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AccuMAP Low pH Protein Digestion Kit enables suppression of sample preparation-induced artificial deamidation, disulfide bond scrambling and oxidation overthe course of sample preparation. Pre-existing non-enzymatic modifications remain intact.
  • Suppress Artificial Deamidation and Disulfide Bond Scrambling in Protein Sample Prep
  • Sample preparation completed in 4.5−5 hours
  • Highly reproducible digestion results
  • Use for reduced and nonreduced proteins
Specifications
Content And Storage <-65°C
Quantity 1 Each