missing translation for 'onlineSavingsMsg'
Learn More

Promega AccuMAP 10X Low pH Reaction Buffer, 1ml

Catalog No. PRVA1010
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRVA1010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRVA1010 Supplier Promega Supplier No. VA1010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AccuMAP Low pH Protein Digestion Kit enables suppression of sample preparation-induced artificial deamidation, disulfide bond scrambling and oxidation overthe course of sample preparation. Pre-existing non-enzymatic modifications remain intact.

  • Suppress Artificial Deamidation and Disulfide Bond Scrambling in Protein Sample Prep
  • Sample preparation completed in 4.5−5 hours
  • Highly reproducible digestion results
  • Use for reduced and nonreduced proteins

Specifications

Content And Storage <-65°C
Quantity 1 mL