Learn More
Accuform Signs Performance Fire Extinguisher Labels
Keep your workforce informed during an emergency and ensure they can quickly locate live-saving equipment with Fire Equipment Labels.
Supplier:  Accuform Signs LFXG420XVE
 View more versions of this product
Catalog No. 19-804-584
Only null left
Description
Description
Specifications
Specifications
Vinyl
3 x 7 in.
Acrylic
-20°C to +80°C
Fire Extinguisher
White, Red
1 Ea.
7.62 x 17.78 cm
6 mil
-20°F to +176°F
Fire Safety Label
5 to 7 Years
Product Suggestions
Product Suggestions