missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Performance Fire Extinguisher Labels

Keep your workforce informed during an emergency and ensure they can quickly locate live-saving equipment with Fire Equipment Labels.

Supplier:  Accuform Signs LFXG519

 View more versions of this product

Catalog No. 19-169-535


Only null left

Description

Description

Specifications

Specifications

Graphic
1 Ea.
5.08 x 12.7 cm
Flammable/combustible
-20°C to +80°C
Use on Flammable Liquids
5 to 7 Years
Vinyl
2 x 5 in.
Acrylic
6 mil
-20°F to +176°F
Fire Safety Label
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions