Learn More
Accuform Signs Performance Fire Extinguisher Labels
Keep your workforce informed during an emergency and ensure they can quickly locate live-saving equipment with Fire Equipment Labels.
Supplier:  Accuform Signs LFXG533
 View more versions of this product
Catalog No. 19-169-530
Only null left
Description
Description
Specifications
Specifications
Graphic
1 Ea.
5.08 x 12.7 cm
Use on ordinary combustibles Paper, wood, cloth, flammable liquids gasoline, Oil, grease, electrical equipment wiring, fuses, circuit breakers
-20°C to +80°C
Use on Class A, B, C Fires
5 to 7 Years
Vinyl
2 x 5 in.
Acrylic
6 mil
-20°F to +176°F
Fire Safety Label
Product Suggestions
Product Suggestions