missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Danger: Confined Space

Colorful, compact safety signs

$17.68 - $58.32

Specifications

Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
Languages English
Displayed Text DANGER Confined Space
View More Specs

Products 4
Catalog Number Mfr. No. Material Height (Metric) Price Quantity  
Catalog Number Mfr. No. Material Height (Metric) Price Quantity  
17-998-082
View Documents
Accuform Signs
MCSP002VA
Aluminum 25.4 cm
Each for $58.32
Only null left
 
17-998-081
View Documents
Accuform Signs
MCSP002VP
Plastic 25.4 cm
Each for $24.70
Only null left
 
17-998-085
View Documents
Accuform Signs
MCSP116VA
Aluminum 17.8 cm
Each for $35.13
Only null left
 
17-998-084
View Documents
Accuform Signs
MCSP116VP
Plastic 17.8 cm
Each for $17.68
Only null left
 
Description

Description

  • Legend: DANGER CONFINED SPACE
  • Available in a variety of materials and sizes
  • Color: White, Black, Red
  • Language: English

Specifications

Specifications

1 Ea.
Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
DANGER Confined Space
Surface Mount
English
Safety Sign
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'