missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Danger: Confined Space

Colorful, compact safety signs

Supplier:  Accuform Signs MCSP116VP

 View more versions of this product

Catalog No. 17-998-084


Only null left
Explore available promotions

Description

Description

  • Legend: DANGER CONFINED SPACE
  • Available in a variety of materials and sizes
  • Color: White, Black, Red
  • Language: English

Specifications

Specifications

Plastic
Surface Mount
7 in.
168°F
0.06 in.
25.4 cm
-90°F to 168°F
DANGER Confined Space
Red on White
1 Ea.
Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
17.8 cm
-90°F
10 in.
-68°C to 76°C
English
Safety Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions