missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Confined Space Do Not Enter
Colorful, compact safety signs
$16.40 - $51.46
Specifications
| Certifications/Compliance | 29 CFR 191.146 |
|---|---|
| Quantity | 1 Ea. |
| For Use With (Application) | Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage |
| Languages | English |
| Displayed Text | DANGER Confined Space Do Not Enter |
| Catalog Number | Mfr. No. | Material | Height (English) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Height (English) | Price | Quantity | ||||
|
17-998-004
|
Accuform Signs
MCSP006VP |
Plastic | 7 in. |
Each for $20.13
|
|
||||
|
17-998-005
|
Accuform Signs
MCSP006VA |
Aluminum | 7 in. |
Each for $30.97
|
|
||||
|
17-998-003
|
Accuform Signs
MCSP006VS |
Vinyl | 7 in. |
Each for $16.40
|
|
||||
|
17-998-002
|
Accuform Signs
MCSP230VA |
Aluminum | 10 in. |
Each for $51.46
|
|
||||
|
17-998-086
|
Accuform Signs
MCSP230VS |
Vinyl | 10 in. |
Each for $21.62
|
|
||||
|
17-998-087
|
Accuform Signs
MCSP230VP |
Plastic | 10 in. |
Each for $24.69
|
|
||||
Description
- Legend: DANGER. CONFINED SPACE. DO NOT ENTER
- Available in a variety of sizes and materials
- Color: White, Black, Red
- Language: English
Specifications
| 29 CFR 191.146 | |
| Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage | |
| DANGER Confined Space Do Not Enter |
| 1 Ea. | |
| English | |
| Safety Sign |