missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Confined Space Do Not Enter
Colorful, compact safety signs
Supplier: Accuform Signs MCSP230VA
Description
- Legend: DANGER. CONFINED SPACE. DO NOT ENTER
- Available in a variety of sizes and materials
- Color: White, Black, Red
- Language: English
Specifications
| 29 CFR 191.146 | |
| 1 Ea. | |
| Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage | |
| 25.4 cm | |
| -40°F | |
| 14 in. | |
| -40°C to 121°C | |
| English | |
| Safety Sign |
| Aluminum | |
| Surface Mount | |
| 10 in. | |
| 250°F | |
| 0.04 in. | |
| 35.6 cm | |
| -40°F to 250°F | |
| DANGER Confined Space Do Not Enter | |
| Black |