Learn More
Thermo Scientific™ Acclaim™ Mixed Mode WCX-1 HPLC Columns
Separate cationic molecules with powerful adjustable selectivity control using this mixed-mode reversed-phase and weak cation-exchange LC column.
$712.86 - $2487.95
Specifications
Pore Size 120 Å
Surface Area 300 m²/g
Temperature 50°C
USP Type L85
Stationary Phase Hydrophobic Alkyl Chain with a Carboxyl Terminus
View More Specs
Products 8
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
DX068352
View Documents
Thermo Scientific™
068352
250 mm 4.6 mm 5 μm
Each for $2,436.18
Only null left
DX068353
View Documents
Thermo Scientific™
068353
150 mm 4.6 mm 5 μm
Each for $2,343.66
Only null left
06-837-1
View Documents
Thermo Scientific™
068371
150 mm 2.1 mm 5 μm
Each for $2,082.53
Only null left
DX071911
View Documents
Thermo Scientific™
071911
10 mm 3 mm 5 μm
Pack of 2 Each for $764.83
Only null left
07-191-0
View Documents
Thermo Scientific™
071910
50 mm 3 mm 3 μm
Each for $1,781.26
Only null left
DX070093
View Documents
Thermo Scientific™
070093
150 mm 2.1 mm 3 μm
Each for $2,487.95
Only null left
DX070092
View Documents
Thermo Scientific™
070092
150 mm 3 mm 3 μm
Each for $2,213.47
Only null left
DX069705
View Documents
Thermo Scientific™
069705
10 mm 4.6 mm 5 μm
Pack of 2 Each for $712.86
Only null left
Description
Description
Separate cationic molecules with powerful adjustable selectivity control using Thermo Scientific™ Acclaim™ Mixed-Mode WCX-1 LC columns. The silica-based stationary phase features a hydrophobic alkyl chain with a carboxyl terminus that provides both hydrophobic reversed-phase and weak cation-exchange properties. This multimode separation mechanism delivers simultaneous separation of basic and neutral compounds. By independently or simultaneously tuning the mobile phase ionic strength, pH, and organic modifier, you can control the selectivity of ionizable and neutral compounds.
Retains and Separates Highly Hydrophilic to Highly Hydrophobic Basic Molecules
  • Multimode retention mechanism: reversed-phase, weak cation-exchange, anion-exclusion, and HILIC modes
  • Selectivity complementary to C18 reversed-phase columns
  • Efficient primary column for a variety of basic analytes; also an ideal column for secondary methods development
  • Versatile in applications that require different selectivities
  • Symmetrical peak shapes and excellent efficiency
  • Stable between pH 2.5 and 7.5
  • Available in 3 and 5μm particle sizes with an average pore diameter of 120Å
Specifications
Specifications
120 Å
50°C
Hydrophobic Alkyl Chain with a Carboxyl Terminus
Mixed Mode
Reversed Phase, Ion Exchange, Ion Exclusion, HILIC
300 m²/g
L85
Acclaim
Proprietary
Responsibly packaged
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.