missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ C30 HPLC Columns
GREENER_CHOICE

Catalog No. DX078666
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Particle Size:
3 μm
5 μm
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
DX078666 50 mm 2.1 mm 3 μm
DX075718 250 mm 4.6 mm 5 μm
DX075719 150 mm 4.6 mm 5 μm
DX078660 100 mm 4.6 mm 3 μm
DX078661 50 mm 4.6 mm 3 μm
DX078664 250 mm 2.1 mm 3 μm
DX078665 100 mm 2.1 mm 3 μm
DX075720 10 mm 4.6 mm 5 μm
DX075721 10 mm 3 mm 5 μm
DX075722 10 mm 2.1 mm 5 μm
DX075723 150 mm 4.6 mm 3 μm
DX075724 150 mm 3 mm 3 μm
DX075725 150 mm 2.1 mm 3 μm
03251446 250 mm 4.6 mm 3 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. DX078666 Supplier Thermo Scientific™ Supplier No. 078666

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for superior separation of hydrophobic structural isomers.

For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.

High Shape Selectivity for the Separation of Structurally Related Isomers
  • Unique selectivity complementary to other reversed-phase columns
  • High efficiency and rugged packing
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 3 and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Å
About the Acclaim Family of HPLC Columns

Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.

  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Flow Rate 0.2 to 0.3 mL/min
Length (Metric) 50 mm
Max. Pressure 4500 psi (310 bar)
Particle Size 3 μm
pH 2 to 8
Pore Size 200 Å
Quantity Each
Surface Area 200 m²/g
Temperature 60°C
USP Type L62
Stationary Phase C30
Product Line Acclaim
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Proprietary
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.