missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ PepMap™ C18 Peptide Analysis HPLC Column, 5 μm, 0.2 mm x 10 mm

Catalog No. DX164212
Change view
Click to view available options
Length (Metric):
10 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
DX164212 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DX164212 Supplier Thermo Scientific™ Supplier No. HSL164212

Column Format: Pre-Column Traps; Carbon Load: 15%; Pore Size: 100 Å; Column Phase: Reversed Phase

Offer high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ Acclaim™ 5μm PepMap™ 100 Nano-Trap Columns.

  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Typically applied for the desalting of peptides before LC separation with MS detection

Specifications

Column Format Pre-Column Traps
Diameter (Metric) 0.2 mm
For Use With LC/MS, ESI-MS, MALDI-MS
Length (Metric) 10 mm
Max. Pressure 14500 psi (1000 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m²/g
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Silica
Carbon Load 15%
Column Type Peptide Analysis
Phase Reversed Phase
Material Fused Silica
Show More Show Less