missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ 300 C18 HPLC Columns

Catalog No. DX069697
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
DX069697 10 mm 4.6 mm
DX060263 50 mm 2.1 mm
DX060264 150 mm 2.1 mm
DX060265 50 mm 4.6 mm
DX060266 150 mm 4.6 mm
DX063684 150 mm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. DX069697 Supplier Thermo Scientific™ Supplier No. 069697

Use these wide-pore C18 columns to achieve high-resolution protein separations.

Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3μm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5μm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.

High-Resolution Separation of Proteins and Peptides
  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1 mL/min
For Use With LC/MS
Length (Metric) 10 mm
Max. Pressure 6000 psi (414 bar)
Particle Size 3 μm
pH 2.5 to 7.5
Pore Size 300 Å
Quantity Pack of 2
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Acclaim
Packing Material Ultrapure Silica
Carbon Load 8%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Technique Bio-LC
Green Features FSC
Product Type Guard Cartridge
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.