Learn More
MilliporeSigma™ Accessories for Samplicity™ Filtration Systems
To be used with the Samplicity™ G2 Filtration System.
Supplier: MilliporeSigma™ SAMP2LID
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.Specifications
| Clear | |
| Samplicity G2 Filtration System with Millex Filters | |
| Analytical sample prep; High Performance Liquid Chromatography (HPLC) | |
| 1 Ea. | |
| Replacement Lid |
| Nonsterile lid with doors for attachment of Millex filters/funnels to HPLC tubes | |
| Thermoplastic Elastomer | |
| PVDF | |
| Up to 8 samples |