Learn More
MilliporeSigma™ Accessories for Samplicity™ Filtration Systems
To be used with the Samplicity™ G2 Filtration System.
Supplier: MilliporeSigma™ SAMTUBING
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.Specifications
| Samplicity G2 Filtration System | |
| Non-sterile | |
| 1 Ea. | |
| Tube Set Assembly |
| Silicone | |
| Analytical sample prep; High Performance Liquid Chromatography (HPLC) | |
| 1 Ea. |