Learn More
Thermo Scientific™ AccelerOme™ TMT11plex MS Sample Preparation Kits
The Thermo Scientific AccelerOme TMT11plex MS Sample Preparation Kit provides a complete solution for efficient and robust proteomics mass spectrometry (MS) sample preparation, including TMT peptide labeling, for use with the AccelerOme Automated Sample Prep System.
$4701.81 - $10655.53
Specifications
Kit Contents Sample prep module, TMT module
Product Type Reagent Kit
Product Line Thermo Scientific
Starting Material Protein Samples
Final Product Type TMT-labeled peptide sample preparation
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Quantity Sufficient For Price Quantity  
PIA50946
View Documents
Thermo Scientific™
A50946
3 x 11 TMT Reactions 10–30 μg samples
Each for $5,021.91
Only null left
PIA50947
View Documents
Thermo Scientific™
A50947
1 x 11 TMT Reactions 30–100 μg samples
Each for $4,701.81
Only null left
PIA50948
View Documents
Thermo Scientific™
A50948
3 x 11 TMT Reactions 30–100 μg samples N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The AccelerOme TMT11plex MS Sample Preparation Kit integrates pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean-up cartridges with TMT11plex label reagents for reproducible sample preparation of TMT-labeled peptides.
Features of the TMT11plex MS Sample Preparation Kit include:
Optimized—EasyPep kit chemistry combined with TMT11plex labeling reagents specifically designed for use with the AccelerOme Automated Sample Prep System
Convenient—complete set of ready-to-use proteomic sample prep, clean-up, and TMT-labeling reagents for easy loading onto the instrument
Robust—reagents combined with AccelerOme ecosystem provide reproducible, high-quality TMT-labeled peptides with greater than 95% labeling efficiency
Flexible—sample prep and TMT-labeling reagents verified on system using cells, plasma, proteins, and tissue samples for 10–100 μg of sample input
Multiplex—enables concurrent mass spectrometry analysis of up to 11 samples
The AccelerOme TMT11plex MS Sample Preparation Kit integrates pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean-up cartridges with TMT11plex label reagents for reproducible sample preparation of TMT-labeled peptides. These single-use components are conveniently provided in labeled trays for direct placement on the AccelerOme system. This kit provides a complete reagent solution for automated sample preparation of TMT11plex labeled samples to produce high quality, mass spectrometry ready peptides.
The AccelerOme reagent portfolio is an integral part of the AccelerOme Automated system solution to provide robust proteomic mass spec sample preparation. The instrument hardware is built on a proven liquid handling format for reproducible sample processing and optimized on-line μSPE sample clean-up for excellent peptide recovery. TMT label reconstitution and sample labeling and pooling have been incorporated before sample clean-up. Peptide quantitation post-clean-up is achieved through accurate UV spectrophotometric analysis to determine peptide concentrations prior to LC-MS analysis. The complete system (kits + reagents + software) was designed to provide high quality peptides while improving productivity with an easy-to-use automated system. When combined with the industry-leading, high-resolution Orbitrap instruments and software, TMT11plex reagents provide integrated total solutions for quantitative protein expression analysis by mass spectrometry.
These products are subject to a limited use label license.
Specifications
Specifications
Sample prep module, TMT module
Thermo Scientific
TMT-labeled peptide sample preparation
Kit
Reagent Kit
Protein Samples
UV Absorption
Automated mass spec sample prep
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'