missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AccelerOme™ System Test Kit

Catalog No. FERA53220
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA53220 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA53220 Supplier Thermo Scientific™ Supplier No. A53220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit provides control reagents for installation and QC of the AccelerOme Automated Sample Prep System.

  • Kit provides control reagents for installation and QC of the AccelerOme Automated Sample Prep System
  • The AccelerOme System Test Kit is required for installation of the AccelerOme Automated Sample Prep System
  • Ready-to-use solutions provided in a tray that is directly inserted into the instrument
  • Buffer matrix and peptide quantitation calibration standard monitor liquid handling and UV spectrophotometer system performance
  • Designed specifically to provide installation and QC support of the AccelerOme system
  • It contains a matrix buffer and peptide quantitation standard to assess the system’s liquid handling reproducibility and accuracy
  • Additionally, the quantitation standard serves as a quality control standard for the on-board UV spectrophotometer
  • The system test kit may also be used for troubleshooting and routine instrument calibration

Specifications

Product Type Reagent Kit
Product Line Thermo Scientific
Starting Material Protein Samples
Detection Method UV Absorption
Quantity 1 Kit