missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Absolute Q™ Liquid Biopsy dPCR Assay - EGFR 6213

Catalog No. A52762
Encompass
Change view
Click to view available options
Absolute Q Liquid Biopsy dPCR Assay - EGFR 6213, 700 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A52762 Absolute Q Liquid Biopsy dPCR Assay - EGFR 6213, 700 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52762 Supplier Applied Biosystems™ Supplier No. A52762
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Absolute Q™ Liquid Biopsy dPCR assays enable reproducible, specific detection of known somatic mutations.

Each predesigned Absolute Q™ Liquid Biopsy dPCR assay has been validated using controls to detect down to 0.1% variant allele frequency in genes relevant for cancer research.

Specifications

Concentration 40X
Detection Method Digital PCR
Format Tube
PCR Method Digital PCR
Product Line Absolute Q
Compatibility QuantStudio™ Absolute Q™ Digital PCR System
Final Product Type Assay
No. of Reactions 700