Learn More
Absolute Q™ Custom FAM dPCR Assays
Catalog No. A53734
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
A53734 1 each
1 options
Catalog No. A53734 Supplier Thermo Fisher Scientific Supplier No. A53734
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Absolute Q™ Custom FAM dPCR Assays
Specifications
Quantity 1 each
How many reactions come in a single order of the Absolute Q Custom FAM dPCR Assay (Cat. No. A53734)?
The Absolute Q Custom FAM dPCR Assay (Cat. No. A53734) comes as a 350 µL volume and can be used in 700 reactions (0.5 µL/reaction).