Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, ROX
Incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
$5,623.04
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
Description
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with ROX or Fluorescein passive reference dyes
Specifications
| 2X | |
| Tube | |
| qPCR | |
| Standard | |
| ABsolute Blue | |
| Gene Expression, Genotyping, siRNA Validation | |
| DNA, dsDNA |
| SYBR | |
| High | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
| Real Time PCR SYBR Master Mix | |
| qPCR Cyclers |
For Research Use Only. Not for use in diagnostic procedures.