missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abiraterone Acetate 98.0+%, TCI America™
Supplier: TCI America A2891200MG
Specifications
| Abiraterone Acetate | |
| Powder/Crystals | |
| MFCD00934213 | |
| 147°C | |
| UVIQSJCZCSLXRZ-UBUQANBQSA-N | |
| [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate | |
| 200 mg | |
| CHEBI:68639 | |
| ≥98.0% (HPLC,T) |
| 154229-18-2 | |
| C26H33NO2 | |
| White/Yellow | |
| 3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate | |
| CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C | |
| 391.555 | |
| 9821849 | |
| 391.56 |
Chemical Identifiers
| 154229-18-2 | |
| 391.555 | |
| UVIQSJCZCSLXRZ-UBUQANBQSA-N | |
| 9821849 | |
| [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
| C26H33NO2 | |
| MFCD00934213 | |
| 3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate | |
| CHEBI:68639 | |
| CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C |
Safety and Handling
RTECSNumber : BV7992100
TSCA : No
Recommended Storage : Freezer