missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse multiclonal Anti-Rat IgG H&L (HRP), 100 uL

Catalog No. 502585498
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502585498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502585498 Supplier Abcam Inc Supplier No. AB106718100UL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Multiclonal antibody, suitable for ELISA, WB.

  • Host Species: Mouse
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: ELISA, WB
  • Species Reactivity: Rat
  • Storage Buffer: Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Western Blot
Conjugate Unconjugated
Formulation Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Multiclonal Antibody
Form Liquid
Isotype IgG
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv ELISA, WB
Show More Show Less