missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse monoclonal [KT100] Anti-Rat IgG+IgM light chain (HRP), 100 uL

Catalog No. 502615314
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502615314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502615314 Supplier Abcam Inc Supplier No. AB106762100UL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal antibody, suitable for ELISA, WB, with 1 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: ELISA, WB
  • Species Reactivity: Rat
  • Storage Buffer: Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ig gamma-1 chain C region
Applications ELISA
Clone KT100
Conjugate Unconjugated
Formulation Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS
Gene Ig gamma-1 chain C region
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Monoclonal Antibody
Isotype IgG2b
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv ELISA, WB
Show More Show Less