missing translation for 'onlineSavingsMsg'
Learn More

ABC Vial™, Sold by MilliporeSigma™ Supelco™

Use with autosamplers that require 12 x 32 mm screw top or crimp top vials. 40% wider opening than standard screw cap vials. Superior thread design ensures a secure seal. Compatible with most GC autosamplers that accept crimp top. Can be used with robotic systems. VialsCaps and septa are not included.

Specifications

Material Glass
Volume (English) 2 mL
View More Specs

Products 6
Catalog Number Mfr. No. Color Marking Spot No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Color Marking Spot No. per Pack Price Quantity & Availability  
111009205
View Documents Promotion Details
N/A Amber Yes 1000 N/A
N/A
111009305
View Documents Promotion Details
N/A Clear Yes 1000 N/A
N/A
111009591
View Documents Promotion Details
N/A Clear No 1000 N/A
N/A
111011787
View Documents Promotion Details
N/A Amber Yes 100 N/A
N/A
111011876
View Documents Promotion Details
N/A Clear Yes 100 N/A
N/A
111011915
View Documents Promotion Details
N/A Clear No 100 N/A
N/A
Description

Description

Autosampler compatibility:

  • Agilent
  • Agilent 1050/1090A
  • Agilent 1100/1200
  • Agilent 7673A, 7683
  • Agilent 5890/7985A/6890
  • Hitachi
  • AS-1000, AS-2000, AS-4000L7200/L7250
  • LEAP Technologies
  • CTC LC PAL, GC PAL
  • CombiPAL
  • HTC PAL/HTS PAL
  • PerkinElmer
  • Autosystem/AS-2000/XL
  • Clarus 500/600
  • ISS-100, 200
  • Shimadzu LC Autosamplers
  • SIL-HTa/SIL-HTc
  • SIL-10ADVP
  • SIL-10A/SIL-10Ai/SIL-10Ap
  • Varian
  • CP-8410
  • 8100/8200
  • 9100/90950/9095
  • Waters
  • Alliance 2690
  • Acquity
  • Breeze
  • 717Plus
Specifications

Specifications

Glass
2 mL
Videos
SDS
Documents

Documents

ABC Vial is a trademark of Wheaton Industries, Inc.