missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AAV-MAX Lysis Buffer

Catalog No. A50520
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50520 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50520 Supplier Gibco™ Supplier No. A50520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells. Cell lysis is detergent-based, eliminating the need for mechanical cell disruption.

Gibco AAV-MAX Lysis Buffer is a ready-to-use, chemically defined cell lysis reagent optimized for adeno-associated virus (AAV) vector production workflows. It uses a gentle, detergent-based formulation to eliminate the need for mechanical cell disruption, providing a milder and easier alternative for isolating AAV particles from HEK293 cell cultures. Supplied as a 10X concentrate, it can be directly added to HEK293 production cultures to induce lysis, streamlining AAV particle extraction and recovery from upstream workflows. AAV-MAX Lysis Buffer integrates seamlessly with the AAV-MAX Production System while remaining compatible with other AAV vector production processes, enabling flexible use across different AAV workflows.

  • Ready-to-use 10X solution—convenient concentrated format can be added directly to cell culture, with no dilution or premixing required
  • Detergent-based cell lysis—gentle chemical lysis avoids mechanical disruption (no sonication or freeze-thaw needed), simplifying AAV particle recovery from cells
  • High-yield AAV extraction—optimized for efficient HEK293 cell lysis for AAV production, maximizing AAV particle recovery and supporting high titers of viral vectors
  • Broad workflow compatibility—functions with the AAV-MAX Production System, from research scale to bioproduction, for versatile application
  • Chemically defined formulation—Polysorbate 20-based, animal-origin free buffer (Triton™ X-100–free) for consistent performance and regulatory compliance

Specifications

Concentration 10X
Product Type Cell Lysis Buffer
Content And Storage 100 mL Bottle, Store at 2°C to 8°C
Shipping Condition Wet Ice
Form Liquid
Product Line AAV-MAX
For Use With (Application) AAV Production
Purification Target AAV
Sufficient For 1 L of Culture
Quantity 100 mL
Composition Polysorbate 20 based
pH 6–8
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.