missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ a-Cyclodextrin

Catalog No. 31801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31801G Supplier R&D Systems™ Supplier No. 3180/1G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Cyclic complexing agent

α-Cyclodextrin (α-CD) is a cylindrical oligosaccharide presenting a hydrophilic surface and a lipophilic cavity. α-Cyclodextrin can form inclusion complexes with a diverse range of hydrophobic compounds. These complexes increase the aqueous solubility, dissolution rate, and bioavailability of the guest compounds.

Specifications

CAS 10016-20-3
Quantity 1 g
Product Line Tocris
Target General Laboratory Research Reagent
Molecular Formula C36H60O30
Purity 0.98