missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ A 867744

Catalog No. 457150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
457150 50 mg
457110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 457150 Supplier R&D Systems™ Supplier No. 4571/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Positive allosteric modulator of α7 nAChR

Positive allosteric modulator of α7 nAChRs (IC50 values are 0.98 and 1.12 μM for human and rat α7 receptor ACh-evoked currents respectively, in X. laevis oocytes). Displays no activity at 5-HT3A, α3β4 or α4β2 nAChRs. Brain penetrant.

Chemical Identifiers

CAS 1000279-69-5
Molecular Formula C20H19ClN2O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 402.89
Purity >97%
Molecular Weight (g/mol) 402.893
Chemical Name or Material A 867744
Recommended Storage Store at +4°C