missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 804598

Catalog No. 447310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
447310 10 mg
447350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 447310 Supplier Tocris Bioscience™ Supplier No. 4473/10

Potent and selective P2X7 antagonist

Potent, competitive P2X7 receptor antagonist (IC50 values are 8.9, 9.9 and 10.9 nM for mouse, rat and human P2X7 receptors respectively). Selective for P2X7 receptors (IC50 > 5-10 μM for a wide array of cell surface receptors and ion channels). Binds with high affinity (Ki app = 2.4 nM for rat P2X7 receptors).

Chemical Identifiers

CAS 1125758-85-1
Molecular Formula C19H17N5

Specifications

Quantity 10 mg
Formula Weight 315.37
Purity >99%
Molecular Weight (g/mol) 315.38
Chemical Name or Material A 804598
Recommended Storage Store at -20°C