missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 286982

Catalog No. 422810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
422810 10 mg
422850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 422810 Supplier Tocris Bioscience™ Supplier No. 4228/10
Only null left

Potent inhibitor of the LFA-1/ICAM-1 interaction

Potent inhibitor of the LFA-1/ICAM-1 interaction (IC50 values are 44 and 35 nM in LFA-1/ICAM-1 binding and LFA-1-mediated cell adhesion assays respectively).

Chemical Identifiers

CAS 280749-17-9
Molecular Formula C24H27N3O4S

Specifications

Quantity 10 mg
Formula Weight 453.55
Purity >99%
Molecular Weight (g/mol) 453.557
Chemical Name or Material A 286982
Recommended Storage Store at Room Temperature