missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 1120

Catalog No. 379310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
379310 10 mg
379350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 379310 Supplier Tocris Bioscience™ Supplier No. 3793/10

High affinity retinol-binding protein 4 (RBP4) ligand

High affinity retinol-binding protein 4 (RBP4) ligand (Ki = 8.3 nM); non-retinoid. Selective against a range of different cellular targets. As efficacious as fenretinide (Cat. No. 1396) in the reduction of serum RBP4 and retinol. Displaces transthyretin (TTR) from RBP4-TTR complexes.

Chemical Identifiers

CAS 1152782-19-8
Molecular Formula C20H19F3N2O3

Specifications

Quantity 10 mg
Formula Weight 392.37
Purity >99%
Molecular Weight (g/mol) 392.378
Chemical Name or Material A 1120
Recommended Storage Store at Room Temperature