missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates

Catalog No. 361023829
Change view
Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Skirt Style:
Skirted
Unskirted
Unskirted, Divisible
Product Type:
Divisible PCR Plates
Low Profile PCR Plates
Microbiology PCR Plates
PCR Plates
Standard PCR Plate
Packaging:
5 Bags x 4 Plates
5 Bags x 5 Plates
Individually Blister Wrapped
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Product Type Packaging
361023829 Colorless Unskirted, Divisible Divisible PCR Plates 5 Bags x 4 Plates
E951020401 Colorless Skirted Standard PCR Plate 5 Bags x 5 Plates
E951020427 Yellow Skirted Standard PCR Plate 5 Bags x 5 Plates
E951020443 Green Skirted Standard PCR Plate 5 Bags x 5 Plates
E951020460 Blue Skirted Standard PCR Plate 5 Bags x 5 Plates
E951020486 Red Skirted Standard PCR Plate 5 Bags x 5 Plates
361023833 Colorless Skirted Microbiology PCR Plates Individually Blister Wrapped
361023834 Blue Skirted Microbiology PCR Plates Individually Blister Wrapped
361023824 Colorless Unskirted Low Profile PCR Plates 5 Bags x 4 Plates
361023827 Blue Unskirted Low Profile PCR Plates 5 Bags x 4 Plates
361023830 Blue Unskirted, Divisible Divisible PCR Plates 5 Bags x 4 Plates
01671100 Colorless Skirted PCR Plates 5 Bags x 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 361023829 Supplier Eppendorf™ Supplier No. 0030133358
Only null left

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

Specifications

Barcode No Barcode
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Colorless
Product Type Divisible PCR Plates
Skirt Style Unskirted, Divisible
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Ideal for extreme applications such as multiple heat sealing
Max. RCF 2250 x g
Profile Low Profile
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Well Color Colorless
Show More Show Less