Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Supplier:  Eppendorf™ 0030113560
 View more versions of this product
Catalog No. 01671100
Only null left
Description
Description
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
Specifications
Chemical Identifiers
Autoclavable
150 μL
Non-sterile
Polypropylene, Polycarbonate
96
Colorless
Specifications
Alphanumeric
Autoclavable
Colorless
Skirted
150 μL
Non-sterile
25 Pk.
Plates with 3-level SafeCode (2D datamatrix, Linear barcode, Alphanumeric code)
5 Bags x 5 Plates
PCR Plates
Polypropylene, Polycarbonate
96
PCR Clean (Non-sterile)
Colorless
Product Suggestions
Product Suggestions