missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Non Binding Microplates

Features long-term surface stability without degradation or leaching.
$1,062.21 - $1,096.75
Specifications
| Format | Standard |
|---|---|
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Polystyrene |
Description
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Significantly increase assay sensitivity
Specifications
| Standard | |
| Not Autoclavable | |
| Polystyrene | |
| Non-sterile | |
| 40/Cs. |
| 8 x 12 | |
| Clear | |
| 96 | |
| 40 | |
| Chimney |
Safety and Handling
ShelfLife : 4 Years