missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Non Binding Microplates

Description
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Significantly increase assay sensitivity
Specifications
Specifications
| Barcode | Alphanumeric |
| Breakapart | No |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Binding Type | Non Binding |
| Color | Clear |
| Product Type | Non-binding Microplate |
| Material | Polystyrene |
| Show More |