missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotix™ 96-Well Format Assay Plates
Large well diameter with raised Chimney-style wells help reduce cross-contamination.
$65.86 - $88.79
Specifications
| Barcode | No Barcode |
|---|---|
| Breakapart | No |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Catalog Number | Mfr. No. | Well Shape | Sterility | Total Volume (Metric) Well | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Well Shape | Sterility | Total Volume (Metric) Well | Price | Quantity & Availability | |||
|
12111070
|
Biotix™
63300102 |
Conical Bottom (V-well) | Non-sterile | 330 μL |
Pack of 10 Each for $65.86
Case of 10 Pack for $658.60
|
|
|||
|
12111071
|
Biotix™
63300103 |
Conical Bottom (V-well) | Sterile | 330 μL |
Pack of 10 Each for $88.79
Case of 10 Pack for $887.90
|
|
|||
Description
- Made from medical grade virgin polypropylene
- DNase, RNase and Endotoxin (pyrogen) free
- Alphanumeric grid to help with sample identification and organization
- Notched bottom corner to be used as a reference for plate orientation
- Available in V-shaped and U-shaped wells
Specifications
| No Barcode | |
| Standard | |
| 8 x 12 | |
| Clear | |
| 96 | |
| 10 | |
| 14.6 mm |
| No | |
| Without Lid | |
| Non Binding | |
| Polypropylene | |
| Non-treated | |
| 85.48 mm |