missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotix™ 96-Well Format Assay Plates
Large well diameter with raised Chimney-style wells help reduce cross-contamination.
Supplier: Biotix™ 63300103
Description
- Made from medical grade virgin polypropylene
- DNase, RNase and Endotoxin (pyrogen) free
- Alphanumeric grid to help with sample identification and organization
- Notched bottom corner to be used as a reference for plate orientation
- Available in V-shaped and U-shaped wells
Chemical Identifiers
| No Barcode | |
| Standard | |
| 8 x 12 | |
| Clear | |
| 96 | |
| 10 | |
| 14.6 mm |
| No | |
| Without Lid | |
| Non Binding | |
| Polypropylene | |
| Non-treated | |
| 85.48 mm |
Specifications
| No Barcode | |
| Standard | |
| 8 x 12 | |
| Clear | |
| Polypropylene | |
| 96 | |
| Non-treated | |
| Conical Bottom (V-well) | |
| 85.48 mm |
| No | |
| Without Lid | |
| Non Binding | |
| Microplate | |
| 330 μL | |
| Sterile | |
| 330 μL | |
| 10 | |
| 14.6 mm |