Learn More
Fisherbrand™ 96-Well DeepWell™ Polypropylene Microplates
GREENER_CHOICE
Ideal for storage because of the low-binding characteristics of polypropylene.
$419.16 - $1,482.41
Specifications
Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
View More Specs
Products 4
Catalog Number Mfr. No. Sterility Volume (Metric) Well Volume (Metric) Working Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Volume (Metric) Well Volume (Metric) Working Price Quantity & Availability  
12566611
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
260254
Sterile 1300 μL 1000 μL
Case of 50 Each for $419.16
Only null left
12566121
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
236800
Non-sterile 2000 μL 1900 μL
Case of 60 Each for $876.03
Only null left
12566612
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
278746
Sterile 2000 μL 1900 μL
Case of 60 Each for $1,482.41
Only null left
12566120
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
236600
Non-sterile 1300 μL 1000 μL
Case of 50 Each for $622.47
Only null left
Description
Description
Ideal for storage because of the low-binding characteristics of polypropylene
  • Accommodate even larger working volumes; up to 1.0 mL or 2.0 mL storage volume per well
  • Proteins and DNA will not adhere to PP, allowing for complete sample recovery.
  • Natural color
  • Nonsterile and sterile available
Specifications
Specifications
Standard
Natural
96
For storage of low-binding characteristics of polypropylene
8 x 12
Polypropylene
Round