missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well DeepWell™ Polypropylene Microplates
GREENER_CHOICE

Catalog No. 12566612
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
1300 μL
2000 μL
Volume (Metric) Working:
1000 μL
1900 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Well Volume (Metric) Working
12566612 Sterile 2000 μL 1900 μL
12566611 Sterile 1300 μL 1000 μL
12566121 Non-sterile 2000 μL 1900 μL
12566120 Non-sterile 1300 μL 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12566612 Supplier Fisherbrand™ Supplier No. 278746
Only null left

Ideal for storage because of the low-binding characteristics of polypropylene.

Ideal for storage because of the low-binding characteristics of polypropylene

  • Accommodate even larger working volumes; up to 1.0 mL or 2.0 mL storage volume per well
  • Proteins and DNA will not adhere to PP, allowing for complete sample recovery.
  • Natural color
  • Nonsterile and sterile available

Specifications

Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Volume (Metric) Working 1900 μL
Sterility Sterile
Well Shape Round
For Use With (Application) For storage of low-binding characteristics of polypropylene