missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Chimney Style, Non-binding Microplates with μClear™ Film Bottom
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well, Chimney Style Non-binding Microplates with Clear™ Film Bottom are free of detectable DNase, RNase, human DNA.
$1564.53 - $1579.03
Specifications
| Breakapart | No |
|---|---|
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Non Binding |
Description
- Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
- Stable under normal assay conditions without degradation or leaching
- Improves signal-to-noise-ratio
- Manufactured under DIN ISO 9001 guidel
Specifications
| No | |
| Without Lid | |
| Non Binding | |
| 96 | |
| Non-treated | |
| Flat Bottom (F-well) | |
| Clear |
| Standard | |
| 8 x 12 | |
| Polystyrene | |
| Non-sterile | |
| 392 μL | |
| 40/Cs. | |
| 127.8 x 85.5 x 14.4 mm |