missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Chimney Style, Non-binding Microplates with μClear™ Film Bottom

Description
- Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
- Stable under normal assay conditions without degradation or leaching
- Improves signal-to-noise-ratio
- Manufactured under DIN ISO 9001 guidelines
Service Plan
No
Warranty
No
Specifications
Specifications
| Barcode | Alphanumeric |
| Breakapart | No |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Non Binding |
| Color | Black, Clear Bottom |
| Product Type | Non-Binding Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 25 to 340 μL |
| Show More |