missing translation for 'onlineSavingsMsg'
Learn More

9-Vinylanthracene, 97%

Catalog No. AAB2233303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2233303 1 g
AAB2233306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2233303 Supplier Thermo Scientific Chemicals Supplier No. B2233303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2444-68-0 | C16H12 | 204.272 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2444-68-0
Molecular Formula C16H12
Molecular Weight (g/mol) 204.272
MDL Number MFCD00003576
InChI Key OGOYZCQQQFAGRI-UHFFFAOYSA-N
Synonym 9-vinylanthracene, anthracene, 9-ethenyl, unii-a3692nge8j, 9-vinyl-anthracene, 9-ethenyl anthracene, 9-ethenylanthracen, 10-vinylanthracene
PubChem CID 17125
IUPAC Name 9-ethenylanthracene
SMILES C=CC1=C2C=CC=CC2=CC3=CC=CC=C31

Specifications

Melting Point 62°C to 65°C
Boiling Point 61°C to 66°C (10 mmHg)
Quantity 1 g
Beilstein 2041641
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 9-Vinylanthracene
EINECSNumber 219-486-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only