accessibility menu, dialog, popup

UserName

CAS RN 2444-68-0

IUPAC Name: 9-ethenylanthracene
Synonyms: 9-ethenylanthracene
Molecular Weight (g/mol): 204.27
Molecular Formula: C16H12
InChi Key: OGOYZCQQQFAGRI-UHFFFAOYSA-N
SMILES: C=CC1=C2C=CC=CC2=CC2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)