missing translation for 'onlineSavingsMsg'
Learn More

9-Trifluoroacetylanthracene 98.0+%, TCI America™

Catalog No. A14225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14225G 5 g
A14221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14225G Supplier TCI America Supplier No. A14225G
Only null left

Chemical Identifiers

CAS 53531-31-0
Molecular Formula C16H9F3O
Molecular Weight (g/mol) 274.242
MDL Number MFCD00001258
InChI Key MNCMBBIFTVWHIP-UHFFFAOYSA-N
Synonym 9-trifluoroacetylanthracene, 9-anthryl trifluoromethyl ketone, 1-anthracen-9-yl-2,2,2-trifluoroethanone, 9-trifluoroacetyl anthracene, 1-9-anthryl-2,2,2-trifluoroethan-1-one, 1-9-anthryl-2,2,2-trifluoroethanone, 9-anthryl, pubchem16290, acmc-209l8i, bidd:gt0598
PubChem CID 104525
IUPAC Name 1-anthracen-9-yl-2,2,2-trifluoroethanone
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 274.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Trifluoroacetylanthracene