CAS RN 53531-31-0
IUPAC Name:
1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-one
Synonyms:
1-(anthracen-9-yl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol):
274.24
Molecular Formula:
C16H9F3O
InChi Key:
MNCMBBIFTVWHIP-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
1
–
1
of
1
results
1
9-Trifluoroacetylanthracene 98.0+%, TCI America™
CAS: 53531-31-0 Molecular Formula: C16H9F3O Molecular Weight (g/mol): 274.242 MDL Number: MFCD00001258 InChI Key: MNCMBBIFTVWHIP-UHFFFAOYSA-N Synonym: 9-trifluoroacetylanthracene,9-anthryl trifluoromethyl ketone,1-anthracen-9-yl-2,2,2-trifluoroethanone,9-trifluoroacetyl anthracene,1-9-anthryl-2,2,2-trifluoroethan-1-one,1-9-anthryl-2,2,2-trifluoroethanone,9-anthryl,pubchem16290,acmc-209l8i,bidd:gt0598 PubChem CID: 104525 IUPAC Name: 1-anthracen-9-yl-2,2,2-trifluoroethanone SMILES: C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F