missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Nitroanthracene 90.0+%, TCI America™
$98.18 - $340.75
Chemical Identifiers
| CAS | 602-60-8 |
|---|---|
| Molecular Formula | C14H9NO2 |
| Molecular Weight (g/mol) | 223.231 |
| MDL Number | MFCD00001248 |
| InChI Key | LSIKFJXEYJIZNB-UHFFFAOYSA-N |
| Synonym | anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene |
| PubChem CID | 11767 |
| ChEBI | CHEBI:82501 |
| IUPAC Name | 9-nitroanthracene |
| SMILES | C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-] |
Chemical Identifiers
| 602-60-8 | |
| 223.231 | |
| LSIKFJXEYJIZNB-UHFFFAOYSA-N | |
| 11767 | |
| 9-nitroanthracene |
| C14H9NO2 | |
| MFCD00001248 | |
| anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene | |
| CHEBI:82501 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-] |
Specifications
| 602-60-8 | |
| 275°C | |
| MFCD00001248 | |
| anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-] | |
| 223.231 | |
| CHEBI:82501 | |
| ≥90.0% (GC) | |
| 9-Nitroanthracene |
| 144°C | |
| C14H9NO2 | |
| 1 g | |
| LSIKFJXEYJIZNB-UHFFFAOYSA-N | |
| 9-nitroanthracene | |
| 11767 | |
| 223.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CB0715000
TSCA : No