accessibility menu, dialog, popup

UserName

CAS RN 602-60-8

IUPAC Name: 9-nitroanthracene
Synonyms: anthracene, 9-nitro-
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: LSIKFJXEYJIZNB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results